Anti-CLCA2 DNA×Pab Rabbit Polyclonal Antibody
Catalog # ABNOH00009635-W01P
Supplier: Abnova
Specifications
- Antigen name:CLCA2 DNA×Pab
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Storage buffer:In 1X PBS, pH 7,4
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Purification:Immunoelectrophoresis (IEP)
- Size:200 μg
- Pk:100 µG
Specifications
About this item
Type:
Antigen: CLCA2 DNAxPab
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human