Order Entry
Switzerland
ContactUsLinkComponent
undefined | Refractive Index Standards
Catalog # REGERIBS40S
Supplier:  REAGECON
CAS Number:  
Catalog # REGERIBS40S
Supplier:  REAGECON
CAS Number:  

Specifications

  • Pack type:
    HDPE dropping bottle
  • Pk:
    15 mL
  • Description:
    Refractive index (stabilised sucrose) standard
  • Storage temperature:
    20 °C
  • Accuracy:
    ±0,00014 nD
  • Refractive index:
    1,39986 nD

Specifications

About this item

Refractive index standards (stabilised sucrose) 1,39986 nD at 20 °C is a high quality product, with an extended shelf life to be used either as a calibrant or analytical control material for use in refractive index based methods for refractive index determination. This product can be used on any make or model of refractometer.

  • Tested using a state of the art refractometer
  • Manufactured gravimetrically and is traceable

It is stable for one year either opened or unopened and is presented in a convenient 15 ml dropper bottle. The product is prepared gravimetrically on a weight/weight basis. Both solute and solvent are weighed on a balance calibrated by OIML traceable weights.

Certifications: ISO17025 accreditation for the test method used to certify this product.