Orders LinkContactUsLinkComponent
Essential Chromatography Pump Systems, C-900 PURE
Essential Chromatography Pump Systems, C-900 PURE
Supplier:   BUCHI
Essential Chromatography Pump Systems, C-900 PURE
Supplier:  BUCHI
Certificates
Essential Chromatography Pump Systems, C-900 PURE
Essential Chromatography Pump Systems, C-900 PURE
4 Options Available Below
Product Details & Documents
C-900 PURE Essential chromatography pump system with flexible software control.
  • Easy accessibility and large touchscreen user interface
  • Basic design and setup
  • Intuitive handling
  • Small footprint and robust
  • Automated capabilities
  • Expansion possibilities
  • No limits for sample loading and fraction collection
The Pure Essential flash chromatography system focuses on the essentials for any basic flash application. The design of the instrument, its modularity, and user-friendliness make it easy for you to take the first steps into automation and profit from increased speed and endless possibilities.
Designed for simplicity, power, and safety, all within a small footprint, the Pure Essential chromatography system is ideal for crowded labs and users taking their first steps into automated chromatography. The instrument’s focus is on getting the core components right and includes a powerful 50 bar pump, an easily configurable UV detector, and a fraction collector that can be added if required.
Supporting you in every step of your research journey, the modular and scalable Pure Essential chromatography system was designed to grow with your research needs. Offering unmatched flexibility and scalability for your separation process.
Delivery information: The scope of delivery depends on the configuration of the purchase order. The pump module includes 1 Pure Chromatography C-900 (pump tower), 1 power cord, 1 solvent line pressure side kit, 2 fitting kits (one-piece fitting ⅛") and 1 operational manual.
Documents
Product Family Options
Product Information
  • Model
    Availability
    Price
  • 11C90000.
    C-900 Pure Essential Chromatography modular system for flash applications, maximum operating pressure (flash mode) 50 bar, flow rate (flash mode) 0 - 300 ml/min., binary gradient flash mode, two solvents
    C-900 Pure Essential Chromatography modular system for flash applications, maximum operating pressure (flash mode) 50 bar, flow rate (flash mode) 0 - 300 ml/min., binary gradient flash mode, two solvents
    C-900
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 6’970.00
    SFr. 6’970.00
    1 * 1 ST
       SFr. 6’970.00
  • 11C90020.
    C-900 Pure Essential Chromatography modular system for flash applications, maximum operating pressure (flash mode) 50 bar, flow rate (flash mode) 0 - 300 ml/min., binary gradient flash mode, two solvents, fraction collector (closed fraction collector bay)
    C-900 Pure Essential Chromatography modular system for flash applications, maximum operating pressure (flash mode) 50 bar, flow rate (flash mode) 0 - 300 ml/min., binary gradient flash mode, two solvents, fraction collector (closed fraction collector bay)
    C-900
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 13’100.00
    SFr. 13’100.00
    1 * 1 ST
       SFr. 13’100.00
  • 11C90030.
    C-900 Pure Essential Chromatography modular system for flash applications, maximum operating pressure (flash mode) 50 bar, flow rate (flash mode) 0 - 300 ml/min., binary gradient flash mode, two solvents, UV detector (254, 275, 325 and 365 nm)
    C-900 Pure Essential Chromatography modular system for flash applications, maximum operating pressure (flash mode) 50 bar, flow rate (flash mode) 0 - 300 ml/min., binary gradient flash mode, two solvents, UV detector (254, 275, 325 and 365 nm)
    C-900
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 12’260.00
    SFr. 12’260.00
    1 * 1 ST
       SFr. 12’260.00
  • 11C90010.
    C-900 Pure Essential Chromatography modular system for flash applications, maximum operating pressure (flash mode) 50 bar, flow rate (flash mode) 0 - 300 ml/min., binary gradient flash mode, two solvents, UV detector (254, 275, 325 and 365 nm), fraction collector (closed fraction collector bay)
    C-900 Pure Essential Chromatography modular system for flash applications, maximum operating pressure (flash mode) 50 bar, flow rate (flash mode) 0 - 300 ml/min., binary gradient flash mode, two solvents, UV detector (254, 275, 325 and 365 nm), fraction collector (closed fraction collector bay)
    C-900
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 17’780.00
    SFr. 17’780.00
    1 * 1 ST
       SFr. 17’780.00
Cartridge holder accessory
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Pivoting clamp for GlasPure ID 15 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - SFr. 117.00
    SFr. 117.00
    1 * 1 Pce
       SFr. 117.00
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Cartridge holder XL for the V-Stand for 3000 g and 5000 g cartridges
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 1’140.00
    SFr. 1’140.00
    1 * 1 ST
       SFr. 1’140.00
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Pivoting clamp for GlasPure ID 70 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 218.00
    SFr. 218.00
    1 * 1 ST
       SFr. 218.00
Product Information
  • Catalog #
    Description
    Availability
    Price
  • V-stand with rod, 950 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 544.00
    SFr. 544.00
    1 * 1 ST
       SFr. 544.00
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Pivoting clamp for GlasPure ID 36 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 155.00
    SFr. 155.00
    1 * 1 ST
       SFr. 155.00
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Cartridge holder XL for the V-Stand for 800 g and 1600 g cartridges
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 933.00
    SFr. 933.00
    1 * 1 ST
       SFr. 933.00
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Pivoting clamp for GlasPure ID 26 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 162.00
    SFr. 162.00
    1 * 1 ST
       SFr. 162.00
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Pivoting clamp for GlasPure ID 100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 394.00
    SFr. 394.00
    1 * 1 ST
       SFr. 394.00
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Pivoting clamp for GlasPure ID 49 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 175.00
    SFr. 175.00
    1 * 1 ST
       SFr. 175.00
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Pure clamp for Pure Essential cartridge stand
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 285.00
    SFr. 285.00
    1 * 1 ST
       SFr. 285.00
Cartridge stand
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Pure Essential cartridge stand for catridges up to 330 g
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 290.00
    SFr. 290.00
    1 * 1 ST
       SFr. 290.00
Connection accessory
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Luer lock connection set
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 59.10
    SFr. 59.10
    1 * 1 ST
       SFr. 59.10
Filter
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Inline filter
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 141.00
    SFr. 141.00
    1 * 1 ST
       SFr. 141.00
Mixing chambers
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Pure mixing chamber, 13 ml volume, for flow rates of 80 - 180 ml/min.
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 323.00
    SFr. 323.00
    1 * 1 ST
       SFr. 323.00
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Pure mixing chamber, 2,5 ml volume, for flow rates of 10 - 35 ml/min.
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 279.00
    SFr. 279.00
    1 * 1 ST
       SFr. 279.00
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Pure mixing chamber, 7 ml volume, for flow rates of 30 - 100 ml/min.
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 296.00
    SFr. 296.00
    1 * 1 ST
       SFr. 296.00
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Pure mixing chamber, 22 ml volume, for flow rates of 130 - 300 ml/min.
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 318.00
    SFr. 318.00
    1 * 1 ST
       SFr. 318.00
Retaining container
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Retaining container for solvent bottle platform for more safety regarding leaking
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 146.00
    SFr. 146.00
    1 * 1 ST
       SFr. 146.00
Sample introduction
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Injection valve UNF ¹/₄"-28 includes T-piece, for convenient injection of up to 5 ml sample
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 185.00
    SFr. 185.00
    1 * 1 ST
       SFr. 185.00
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Six-way valve for safe and fast sample loading
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 1’960.00
    SFr. 1’960.00
    1 * 1 ST
       SFr. 1’960.00
Sample introduction accessories
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Sample loop for sample volumes of 1 - 20 ml, FEP coil
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 311.00
    SFr. 311.00
    1 * 1 ST
       SFr. 311.00
Product Information
  • Catalog #
    Description
    Availability
    Price
  • 500 ml glass part for sample chamber 100 ml, for convenient injection of samples volumes up to 500 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 638.00
    SFr. 638.00
    1 * 1 ST
       SFr. 638.00
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Sample loop for sample volumes of 1 - 5 ml, FEP coil
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 224.00
    SFr. 224.00
    1 * 1 ST
       SFr. 224.00
Product Information
  • Catalog #
    Description
    Availability
    Price
  • 250 ml glass part for sample chamber 100 ml, for convenient injection of samples volumes up to 250 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 579.00
    SFr. 579.00
    1 * 1 ST
       SFr. 579.00
Product Information
  • Catalog #
    Description
    Availability
    Price
  • PURE solid loader M set, incl. adapter set, sleeve, tubes (20 pieces) and frits (40 pieces)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 1’460.00
    SFr. 1’460.00
    1 * 1 ST
       SFr. 1’460.00
Product Information
  • Catalog #
    Description
    Availability
    Price
  • PURE solid loader S set, incl. adapter set, sleeve, tubes (20 pieces) and frits (40 pieces)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 863.00
    SFr. 863.00
    1 * 1 ST
       SFr. 863.00
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Sample chamber 100 ml set includes all adapters needed for operation, for convenient injection of samples volumes between 10 - 100 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 877.00
    SFr. 877.00
    1 * 1 ST
       SFr. 877.00
Product Information
  • Catalog #
    Description
    Availability
    Price
  • 1000 ml glass part for sample chamber 100 ml, for convenient injection of samples volumes up to 1000 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 755.00
    SFr. 755.00
    1 * 1 ST
       SFr. 755.00