Aluminized Neck Protection
JUTEHSN030KA-1
: JUTEC
- Garment application:Industrial
- Risk area:Heat
- Length:300 mm
- Pk:1
Neck protection made of preox-aramide fabric.
- Fabric resistant to flying sparks
- Flexible, lightweight and soft
- Including Velcro fastener
- Suitable for Schuberth and Voss helmets
- Universal dimensions
- Weight: ~260 g/m²
: EN ISO 11612:2015.