ContactUsLinkComponent
FlashPure EcoFlex Diol Cartridges
 :  BUCHI
FlashPure EcoFlex Diol Cartridges
 :  BUCHI
  • Catalog No:
  • 140000074.
  • 140000072.
  • 140000073.
  • 140000067.
  • 140000070.
  • 140000071.
  • 140000068.
  • 140000069.
  • Phase name:
  • FlashPure EcoFlex diol
  • FlashPure EcoFlex diol
  • FlashPure EcoFlex diol
  • FlashPure EcoFlex diol
  • FlashPure EcoFlex diol
  • FlashPure EcoFlex diol
  • FlashPure EcoFlex diol
  • FlashPure EcoFlex diol
  • Carbon load:
  • ≥4%
  • ≥4%
  • ≥4%
  • ≥4%
  • ≥4%
  • ≥4%
  • ≥4%
  • ≥4%
  • Endcapped:
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • Max. pressure:
  • 12 bar
  • 12 bar
  • 12 bar
  • 12 bar
  • 12 bar
  • 12 bar
  • 12 bar
  • 12 bar
  • Surface area:
  • 260 - 360 m²/g
  • 260 - 360 m²/g
  • 260 - 360 m²/g
  • 260 - 360 m²/g
  • 260 - 360 m²/g
  • 260 - 360 m²/g
  • 260 - 360 m²/g
  • 260 - 360 m²/g
  • Polarity:
  • Mid-polarity stationary phase
  • Mid-polarity stationary phase
  • Mid-polarity stationary phase
  • Mid-polarity stationary phase
  • Mid-polarity stationary phase
  • Mid-polarity stationary phase
  • Mid-polarity stationary phase
  • Mid-polarity stationary phase
Frequently Bought Together