ContactUsLinkComponent
MELAcontrol Pro for Stream Sterilizers
MELAcontrol Pro for Stream Sterilizers
Catalog # MELAME01075
MELAcontrol Pro for Stream Sterilizers
Catalog # MELAME01075
Specifications
  • Description:
    MELAcontrol Pro
  • Pk:
    1
Specifications
About this item
MELAcontrol Pro with 40 indicator strips is a durable, usable test body for the power-user.
This is a test body for 'Class B' steam sterilizers, which simulate the complicated requirements for sterilizing hollow instruments. This test body is required for batch monitoring of the processing of 'critical B' instruments.
Certifications: EN 867-5 standard.
Ordering information: The test strips can be re-ordered in a refill package with 250 indicator strips.
Disposable paper filter
Product Information