Advanced TC™ cell culture flasks
About this item
Cell culture flasks with a standard or ventilated cap and a modified culture surface to optimise cell characteristics and functions. The improved adherence and increased growth rate improve and accelerate cell proliferation. Additionally, the Advanced TC™ surface facilitates uniform and consistent cell adherence, increases throughput and reduces cell loss, for example during automated washing stages. Free of detectable DNase, RNase and human DNA. Free of cytotoxic substances.
- Standard or ventilated cap
- Increased cell adherence
- Uniform and consistent cell adherence
- Consistent cell growth
- Morphology similar to in vivo conditions
- Increased throughput
- Optimal cultivation conditions for sensitive cells
2 Options Available Below
Product Details & Documents
Cell culture flasks with a standard or ventilated cap and a modified culture surface to optimise cell characteristics and functions. The improved adherence and increased growth rate improve and accelerate cell proliferation. Additionally, the Advanced TC™ surface facilitates uniform and consistent cell adherence, increases throughput and reduces cell loss, for example during automated washing stages. Free of detectable DNase, RNase and human DNA. Free of cytotoxic substances.
- Standard or ventilated cap
- Increased cell adherence
- Uniform and consistent cell adherence
- Consistent cell growth
- Morphology similar to in vivo conditions
- Increased throughput
- Optimal cultivation conditions for sensitive cells
Specifications for Product Family
Specifications
- Catalog No:
- 392-0234
- 392-0236
- 392-0237
- 392-0240
- 392-0241
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
Specifications
Product Family Options
Product Information
- CapacityGrowth areaClosure typeRecommended working volumeAvailabilityPrice
- Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.392-0234Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.392-0236Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact - 550 ml175 cm²Filter cap20 - 45 mlDiscontinued-Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.392-0237Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact - 650 ml175 cm²Standard cap20 - 85 mlDiscontinued-Specifications:
More
Specifications
Cat. No.392-0240Environmentally PreferableEnvironmental certification - 650 ml175 cm²Filter cap20 - 85 mlDiscontinued-Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.392-0241Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact