PEG ELISA kit
ABNOKA6892.1KIT
: Abnova
:
- Pk:1 KIT
- Target protein:PEG
PEG ELISA Kit used an antibody that recognizes the polyoxyethylene backbone of PEG. Sensitivity varies with PEG molecular weight.
PEG ELISA Kit used an antibody that recognizes the polyoxyethylene backbone of PEG. Sensitivity varies with PEG molecular weight.