Order Entry
Switzerland
ContactUsLinkComponent
Monkey Anti-PEG IgG ELISA kit
Monkey Anti-PEG IgG ELISA kit
Catalog # ABNOKA6898.1KIT
Supplier:  Abnova
CAS Number:  
Monkey Anti-PEG IgG ELISA kit
Catalog # ABNOKA6898.1KIT
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Monkey
  • Target protein:
    Anti-PEG IgG

Specifications

About this item

Monkey Anti-PEG IgG ELISA kit can be used for detection of anti-PEG IgG in monkey serum, plasma and tissue fluids.