Human Enterovirus 71 Antigen ELISA kit
Catalog # ABNOKA6890.1KIT
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Human
- Target protein:Enterovirus 71 Antigen
- Assay Principle:Qualitative
Specifications
About this item
Human Enterovirus 71 Antigen ELISA Kit is an Enzyme-Linked Immunosorbent Assay for the qualitative detection of Human Enterovirus 71 Antigen in serum, plasma, cell culture supernates, cell lysates, tissue homogenates.