Order Entry
Switzerland
ContactUsLinkComponent
Avelumab (Human) ELISA kit
Avelumab (Human) ELISA kit
Catalog # ABNOKA6832.1KIT
Supplier:  Abnova
CAS Number:  
Avelumab (Human) ELISA kit
Catalog # ABNOKA6832.1KIT
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Human
  • Target protein:
    Avelumab
  • Assay Principle:
    Quantitative

Specifications

About this item

Avelumab (Human) Elisa Kit (Quantitative) is a quantitative determination of free Avelumab in serum and plasma.