Order Entry
Switzerland
ContactUsLinkComponent
 
The IgA (Monkey) ELISA kit
Catalog # ABNOKA6755.1KIT
Supplier:  Abnova
CAS Number:  
The IgA (Monkey) ELISA kit
Catalog # ABNOKA6755.1KIT
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Monkey
  • Target protein:
    IgA

Specifications

About this item

The IgA (Monkey) ELISA Kit is used for immunoperoxidase Assay for Determination of IgA in Monkey Samples.