Order Entry
Switzerland
ContactUsLinkComponent
The IgG1 (Monkey) ELISA kit
The IgG1 (Monkey) ELISA kit
Catalog # ABNOKA6750.1KIT
Supplier:  Abnova
CAS Number:  
The IgG1 (Monkey) ELISA kit
Catalog # ABNOKA6750.1KIT
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Monkey
  • Target protein:
    IgG1

Specifications

About this item

The IgG1 (Monkey) ELISA Kit is used for determination of IgG1 in old world monkey serum or plasma.