Auftragserfassung
Switzerland
ContactUsLinkComponent
The IgG1 (Rat) ELISA kit
The IgG1 (Rat) ELISA kit
Catalog # ABNOKA6752.1KIT
Supplier:  Abnova
CAS Number:  
The IgG1 (Rat) ELISA kit
Catalog # ABNOKA6752.1KIT
Supplier:  Abnova
CAS Number:  

Specifications

  • VE:
    1 KIT
  • Reaktivität der primären Antikörper:
    Rat
  • Target protein:
    IgG1

Specifications

About this item

The IgG1 (Rat) ELISA Kit is used for determination of IgG1 in rat serum or plasma.