IgG2b (Rat) ELISA kit
Catalog # ABNOKA6760.1KIT
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Rat
- Target protein:IgG2b
- Assay Principle:Quantitative
Specifications
About this item
IgG2b (Rat) ELISA Kit is for the quantitative measurement of rat IgG2a in serum or plasma.