Order Entry
Switzerland
ContactUsLinkComponent
IgG (Cow) ELISA kit
IgG (Cow) ELISA kit
Catalog # ABNOKA6762.1KIT
Supplier:  Abnova
CAS Number:  
IgG (Cow) ELISA kit
Catalog # ABNOKA6762.1KIT
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Bovine
  • Target protein:
    IgG
  • Assay Principle:
    Quantitative

Specifications

About this item

IgG (Cow) ELISA Kit is for the quantitative measurement of cow IgG in serum, plasma or milk.