IgM (Goat) ELISA kit
ABNOKA6769.1KIT
: Abnova
:
- Pk:1 KIT
- Primary antibody reactivity:Goat
- Target protein:IgM
- Assay Principle:Quantitative
IgM (Goat) ELISA Kit is for the quantitative measurement of goat IgM in serum or plasma.
IgM (Goat) ELISA Kit is for the quantitative measurement of goat IgM in serum or plasma.