Order Entry
Switzerland
ContactUsLinkComponent
IgG (Camel) ELISA kit
IgG (Camel) ELISA kit
Catalog # ABNOKA6770.1KIT
Supplier:  Abnova
CAS Number:  
IgG (Camel) ELISA kit
Catalog # ABNOKA6770.1KIT
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Camel
  • Target protein:
    IgG
  • Assay Principle:
    Quantitative

Specifications

About this item

IgG (Camel) ELISA Kit is for the quantitative measurement of camel IgG in serum or plasma.