Measles Virus IgG (Human) ELISA kit
Catalog # ABNOKA6781.1KIT
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Human
- Target protein:IgG
- Assay Principle:Qualitative
Specifications
About this item
Measles Virus IgG (Human) ELISA Kit is a qualitative determination of IgG class antibodies against Measles Virus in human serum or plasma.