Order Entry
Switzerland
ContactUsLinkComponent
The TNP (Mouse) IgM ELISA kit
The TNP (Mouse) IgM ELISA kit
Catalog # ABNOKA6744.1KIT
Supplier:  Abnova
CAS Number:  
The TNP (Mouse) IgM ELISA kit
Catalog # ABNOKA6744.1KIT
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    TNP

Specifications

About this item

The TNP (Mouse) IgM ELISA Kit is based on a solid phase enzymelinked immunosorbent assay (ELISA). The assay uses TNP-BSA as the capture antigen (coated on the microtiter wells) and HRP conjugated anti-mouse IgM antibodies for detection.