Epstein-Barr Virus EBNA-1 IgG ELISA kit
Catalog # ABNOKA6728.1KIT
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 KIT
- Target protein:IgG
- Assay Principle:Qualitative
Specifications
About this item
Epstein-Barr Virus EBNA-1 IgG ELISA Kit is a qualitative determination of IgG class antibodies against Epstein-Barr virus nuclear antigen type 1 (EBV-EBNA-1) in human serum or plasma.