Epstein-Barr Virus (VCA) IgG ELISA kit
Catalog # ABNOKA6726.1KIT
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Epstein Barr virus
- Target protein:IgG
- Assay Principle:Qualitative
Specifications
About this item
Epstein-Barr Virus (VCA) IgG ELISA Kit is a qualitative determination of IgG class antibodies against Epstein-Barr Virus (VCA) in human serum or plasma.