Aspergillus fumigatus IgG ELISA kit
Catalog # ABNOKA6717.1KIT
Anbieter: Abnova
CAS-Nummer:
Technische Daten
- Pk:1 KIT
- Primary antibody reactivity:Aspergillus fumigatus
- Target protein:IgG
- Assay Principle:Qualitative
Technische Daten
Über diesen Artikel
Aspergillus fumigatus IgG ELISA Kit is a qualitative determination of IgM class antibodies against Aspergillus fumigatus in human serum or plasma.