Toxoplasma IgG ELISA kit
Catalog # ABNOKA6700.1KIT
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Toxoplasma gondii
- Target protein:IgG
- Assay Principle:Quantitative
Specifications
About this item
Toxoplasma IgG ELISA Kit is enzyme immunoassay for the quantitative and qualitative measurement of IgG antibodies to Toxoplasma gondii in human serum and plasma.