FSH (Human) ELISA kit
ABNOKA6704.1KIT
: Abnova
:
- Pk:1 KIT
- Primary antibody reactivity:Human
- Target protein:FSH
- Assay Principle:Quantitative
FSH (Human) ELISA Kit is an enzyme immunoassay for the quantitative measurement of follicle-stimulating hormone in serum.