HSV-1 IgM ELISA kit
Catalog # ABNOKA6712.1KIT
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 KIT
- Target protein:HSV-1 IgM
- Assay Principle:Quantitative
Specifications
About this item
HSV-1 IgM ELISA Kit is an enzyme immunoassay for the qualitative and semiquantitative measurement of IgM antibody to the HSV-1 in human serum and plasma.