Recombinant Human PDGFC (from E. coli), His-tag
Catalog # ABNOP9015.20UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:20 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:His-tag
- Gene ID:Q9NRA1
- Protein/peptide name:PDGFC
- Sequence:235a.a.-345a.a.