Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Recombinant Fish PRLR (from E. coli)
Recombinant Fish PRLR (from E. coli)
Recombinant Fish PRLR (from E. coli)
ABNOP9059.20UG
:
Abnova
:
Recombinant Fish PRLR (from E. coli)
ABNOP9059.20UG
:
Abnova
:
Pk:
20 µG
Protein/peptide type:
Recombinant
Source:
E. coli
Species:
Fish
Gene ID:
Q9DE08
Protein/peptide name:
PRLR