Order Entry
Switzerland
ContactUsLinkComponent
Recombinant Vaccinia virus capping enzyme (from E. coli)
Catalog # ABNOP9373.50UL
Anbieter:  Abnova
CAS-Nummer:  
Recombinant Vaccinia virus capping enzyme (from E. coli)
Catalog # ABNOP9373.50UL
Anbieter:  Abnova
CAS-Nummer:  

Technische Daten

  • Pk:
    50 µl
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Enzyme name:
    Vaccinia virus capping enzyme

Technische Daten

Über diesen Artikel