SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Enzymes
Recombinant Vaccinia virus capping enzyme (from E. coli)
Recombinant Vaccinia virus capping enzyme (from E. coli)
Catalog # ABNOP9375.1ML
Anbieter:
Abnova
CAS-Nummer:
Expand image
Recombinant Vaccinia virus capping enzyme (from E. coli)
Catalog # ABNOP9375.1ML
Anbieter:
Abnova
CAS-Nummer:
Technische Daten
Pk:
1 mL
Enzyme type:
Recombinant
Source:
E. coli
Enzyme name:
Vaccinia virus capping enzyme
Technische Daten
Über diesen Artikel
Product Details
Product Details