Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Recombinant Poly(A) Polymerase (from E. coli)

Recombinant Poly(A) Polymerase (from E. coli)

20 µl
Catalog # ABNOP9379.20UL
Supplier:   Abnova
Recombinant Poly(A) Polymerase (from E. coli)
Catalog # ABNOP9379.20UL
Supplier:  Abnova
CAS Number:  
Recombinant Poly(A) Polymerase (from E. coli)

20 µl

CAS Number:  
Price
559.00 SFr.
Log in for availability
1 * 20 µl
Product Details & Documents
Specifications

Specifications

  • Pk:
    20 µl
  • Cat. No.:
    ABNOP9379.20UL
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Enzyme name:
    Poly(A) Polymerase

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.