Recombinant Mouse DHH (from E. coli)
25 µG
Catalog # ABNOP9139.25UG
Supplier: Abnova
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:25 µG
- Cat. No.:ABNOP9139.25UG
- Protein/peptide type:Recombinant
- Protein/peptide name:DHH
- Species:Mouse
- Source:E. coli
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



