Order Entry
Switzerland
ContactUsLinkComponent
Recombinant Bovine ENTEROKINASE (from P. pastoris), His-tag
Recombinant Bovine ENTEROKINASE (from P. pastoris), His-tag
Catalog # ABNOP7541.10UG
Supplier:  Abnova
CAS Number:  
Recombinant Bovine ENTEROKINASE (from P. pastoris), His-tag
Catalog # ABNOP7541.10UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Enzyme type:
    Recombinant
  • Source:
    P. pastoris
  • Species:
    Bovine
  • Tag sequence:
    His-tag
  • Enzyme name:
    Enterokinase

Specifications

About this item