Anti-CD38 Rabbit Recombinant Antibody [clone: 6F10]
Catalog # ABNORAB04000.100UL
Supplier: Abnova
Specifications
- Antigen symbol:CD38
- Clonality:Recombinant
- Clone:6F10
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Immunogen:Recombinant rabbit monoclonal antibody raised against human CD38.
- Pk:100 µl