Anti-MYC Mouse Recombinant Antibody [clone: 9E10]
Catalog # ABNORAB03383.200UG
Supplier: Abnova
Specifications
- Antigen symbol:MYC
- Clonality:Recombinant
- Clone:9E10
- Host:Mouse
- Isotype:IgG2c kappa
- Reactivity:Human
- Environmentally Preferable:
- Immunogen:Recombinant mouse monoclonal antibody raised against human MYC.
- Pk:200 µG