Anti-ADRB2 Rabbit Recombinant Antibody [clone: HMBT-6B2]
ABNORAB03108.200UG
: Abnova
- Antigen-Symbol:ADRB2
- Klonalität:Recombinant
- Klon:HMBT-6B2
- Wirt:Rabbit
- Isotyp:
- Reaktivität:Mouse
- Environmentally Preferable:
- Immunogen:Recombinant rabbit monoclonal antibody raised against mouse ADRB2.
- VE:200 µG