Anti-TNFRSF9 Rabbit Recombinant Antibody [clone: 4B4-1-1]
Catalog # ABNORAB03646.200UG
Supplier: Abnova
Specifications
- Antigen symbol:TNFRSF9
- Clonality:Recombinant
- Clone:4B4-1-1
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Immunogen:Recombinant rabbit monoclonal antibody raised against human TNFRSF9.
- Pk:200 µG