Anti-TNFRSF9 Rat Recombinant Antibody [clone: TKS-1]
ABNORAB03652.200UG
: Abnova
- Antigen symbol:TNFRSF9
- Clonality:Recombinant
- Clone:TKS-1
- Host:Rat
- Isotype:
- Reactivity:Mouse
- Environmentally Preferable:
- Immunogen:Recombinant rat monoclonal antibody raised against mouse Tnfsf9.
- Pk:200 µG