Anti-USP11 Rabbit Recombinant Antibody [clone: R08-2D7]
ABNORAB01679.100UL
: Abnova
- Antigen symbol:USP11
- Clonality:Recombinant
- Clone:R08-2D7
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Immunogen:Recombinant rabbit monoclonal antibody raised against synthetic peptide of human USP11
- Pk:100 µl