Anti-IKK alpha Rabbit Recombinant Antibody [clone: R03-1E7]
ABNORAB01683.100UL
: Abnova
- Antigen symbol:IKK alpha
- Clonality:Recombinant
- Clone:R03-1E7
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Immunogen:Recombinant rabbit monoclonal antibody raised against synthetic peptide of human IKK alpha
- Pk:100 µl