Anti-Glypican 1/GPC1 Rabbit Recombinant Antibody [clone: R03-2E8]
ABNORAB01691.100UL
: Abnova
- Antigen symbol:Glypican 1/GPC1
- Clonality:Recombinant
- Clone:R03-2E8
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Immunogen:Recombinant rabbit monoclonal antibody raised against synthetic peptide of human Glypican 1/GPC1
- Pk:100 µl