Anti-CD90 Rabbit Recombinant Antibody [clone: R09-6G7]
Catalog # ABNORAB01700.100UL
Supplier: Abnova
Specifications
- Antigen symbol:CD90
- Clonality:Recombinant
- Clone:R09-6G7
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Immunogen:Recombinant rabbit monoclonal antibody raised against synthetic peptide of human CD90
- Pk:100 µl