Anti-Chk2 Rabbit Recombinant Antibody [clone: R06-1B4]
ABNORAB01504.100UL
: Abnova
- Antigen symbol:Chk2
- Clonality:Recombinant
- Clone:R06-1B4
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Immunogen:Recombinant rabbit monoclonal antibody raised against human Chk2.
- Pk:100 µl