Order Entry
Switzerland
ContactUsLinkComponent
Anti-CDT2 Rabbit Recombinant Antibody [clone: R03-2G4]
Anti-CDT2 Rabbit Recombinant Antibody [clone: R03-2G4]
Catalog # ABNORAB01507.100UL
Supplier:  Abnova
Anti-CDT2 Rabbit Recombinant Antibody [clone: R03-2G4]
Catalog # ABNORAB01507.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    CDT2
  • Clonality:
    Recombinant
  • Clone:
    R03-2G4
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human CDT2.
  • Pk:
    100 µl

Specifications

About this item