Order Entry
Switzerland
ContactUsLinkComponent
Anti-CD32 Rabbit Recombinant Antibody [clone: R09-5D1]
Anti-CD32 Rabbit Recombinant Antibody [clone: R09-5D1]
Catalog # ABNORAB01524.100UL
Supplier:  Abnova
Anti-CD32 Rabbit Recombinant Antibody [clone: R09-5D1]
Catalog # ABNORAB01524.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    CD32
  • Clonality:
    Recombinant
  • Clone:
    R09-5D1
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against synthetic peptide of human CD32.
  • Pk:
    100 µl

Specifications

About this item