Order Entry
Switzerland
ContactUsLinkComponent
Anti-CDC37 Rabbit Recombinant Antibody [clone: R04-4I8]
Anti-CDC37 Rabbit Recombinant Antibody [clone: R04-4I8]
Catalog # ABNORAB01794.100UL
Supplier:  Abnova
Anti-CDC37 Rabbit Recombinant Antibody [clone: R04-4I8]
Catalog # ABNORAB01794.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    CDC37
  • Clonality:
    Recombinant
  • Clone:
    R04-4I8
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human CDC37.
  • Pk:
    100 µl

Specifications

About this item