Anti-ALDH1A1 Rabbit Recombinant Antibody [clone: R09-3B9]
Catalog # ABNORAB01301.100UL
Supplier: Abnova
Specifications
- Antigen symbol:ALDH1A1
- Clonality:Recombinant
- Clone:R09-3B9
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Immunogen:Recombinant rabbit monoclonal antibody raised against human ALDH1A1.
- Pk:100 µl