Anti-SRSF3 Rabbit Recombinant Antibody [clone: R01-6G2]
Katalognr. ABNORAB01303.100UL
Anbieter: Abnova
Technische Daten
- Antigen symbol:SRSF3
- Clonality:Recombinant
- Clone:R01-6G2
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Immunogen:Recombinant rabbit monoclonal antibody raised against human SRSF3.
- Pk:100 µl