Order Entry
Switzerland
ContactUsLinkComponent
Anti-MCM2 Rabbit Recombinant Antibody [clone: R03-1B3]
Anti-MCM2 Rabbit Recombinant Antibody [clone: R03-1B3]
Catalog # ABNORAB01781.100UL
Supplier:  Abnova
Anti-MCM2 Rabbit Recombinant Antibody [clone: R03-1B3]
Catalog # ABNORAB01781.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    MCM2
  • Clonality:
    Recombinant
  • Clone:
    R03-1B3
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human MCM2.
  • Pk:
    100 µl

Specifications

About this item