Anti-EIF4EBP1 Rabbit Recombinant Antibody [clone: R03-5C7]
ABNORAB01233.100UL
: Abnova
- Antigen symbol:EIF4EBP1
- Clonality:Recombinant
- Clone:R03-5C7
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Immunogen:Recombinant rabbit monoclonal antibody raised against human EIF4EBP1.
- Pk:100 µl